Learn More
Meso Scale Discovery V-PLEX Plus Rat IFN-y Kit
SDP
Supplier:  Meso Scale Discovery K153QOG2
Encompass
The V-PLEX Plus Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for quantifying rat interferon gamma (IFN-y) in various samples It includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and plate seals The assay s sensitivity is 0 71 pg/mL with a range of 0 71-2500 pg/mL using MSD s SECTOR Imager for detection with 25 L of sample It offers high specificity minimal cross-reactivity and reproducible results with precision of 10%
Catalog No. 50-000-81464