Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-13 Kit
SDP
Supplier:  Meso Scale Discovery K153ODG4
Encompass
The Meso Scale Discovery V-PLEX Plus Rat IL-13 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-13 in rat serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 53-2500 pg/mL and takes approximately 3 5 hours to complete It uses an instrument such as the MSD QuickPlex SQ 120 for plate reading The kit provides materials for 1 x 96-well plate and can assay 40 samples in duplicate
Catalog No. 50-000-82257
Add to Cart
Request a Quote