Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-4 Kit
SDP
Supplier:  Meso Scale Discovery K153QRG2
Encompass
The V-PLEX Plus Rat IL-4 Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of rat interleukin 4 (IL-4) in cell culture supernatants serum plasma and tissue lysates The kit contains a 96-well plate coated with a rat IL-4 capture antibody a ruthenium-labeled rat IL-4 detection antibody calibrators and buffers for sample dilution The assay has a sensitivity of 0 3 pg/mL and a detection range of 0 3-2500 pg/mL It requires 2 5 hours to complete and can detect rat IL-4 with high specificity and sensitivity without interference from other cytokines The kit includes all necessary reagents and can analyze 46 samples in duplicate
Catalog No. 50-000-81170