Learn More
Meso Scale Discovery V-PLEX Plus Rat IL-6 Kit
SDP
Supplier:  Meso Scale Discovery K153QXG4
Encompass
The V-PLEX Plus Rat IL-6 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-6 in rat serum plasma and cell culture supernatants The kit includes a V-PLEX Rat IL-6 Plate that is coated with a capture antibody specific for rat IL-6 The kit also contains a V-PLEX Rat IL-6 Detection Antibody that is conjugated to an ECL label The assay is performed by adding samples and detection antibody to the plate washing then adding read buffer before analyzing on an MSD instrument The sensitivity of the assay is 1 3 pg/mL with an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-78345
Only null left
Add to Cart
Request a Quote