Learn More
Meso Scale Discovery V-PLEX Plus Rat KC/GRO Kit
SDP
Supplier:  Meso Scale Discovery K153QTG4
Encompass
The V-PLEX Plus Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Diluent and lyophilized KC and GRO standards to generate standard curves and quantify sample levels This electrochemiluminescent assay provides high sensitivity and a broad dynamic range The kit can test up to 80 samples in duplicate and has a testing time of 3 5 hours
Catalog No. 50-000-81562
Only null left
Add to Cart
Request a Quote