Learn More
Meso Scale Discovery V-PLEX Rat IFN-y Kit
SDP
Supplier:  Meso Scale Discovery K153QOD4
Encompass
The V-PLEX Rat IFN-y Kit from Meso Scale Discovery is an electrochemiluminescent immunoassay for the quantitative measurement of interferon gamma (IFN-y) in rat serum plasma and cell culture supernatants The assay utilizes MSD s proprietary V-PLEX technology is highly sensitive has a broad dynamic range of up to 3 logs and shows minimal cross-reactivity The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve Sample incubation time is just 2 hours The assay can detect IFN-y concentrations as low as 1 3 pg/mL
Catalog No. 50-000-78346
Add to Cart
Request a Quote