Learn More
Meso Scale Discovery V-PLEX Rat IL-10 Kit
SDP
Supplier:  Meso Scale Discovery K153QUD4
Encompass
The V-PLEX Rat IL-10 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of interleukin 10 (IL-10) in rat serum plasma and cell culture supernatants The assay has high sensitivity a broad dynamic range and requires only 50 L sample size The kit includes a 96-well plate coated with IL-10 capture antibody detection antibody conjugated to an ECL label calibrators diluent wash buffer read buffer and plate seals The assay takes 3 5 hours to complete with only 30 minutes hands-on time and has a lower limit of detection of 1 27 pg/mL
Catalog No. 50-000-81461
Add to Cart
Request a Quote