Learn More
Meso Scale Discovery V-PLEX Rat IL-13 Kit
SDP
Supplier:  Meso Scale Discovery K153ODD1
Encompass
The V-PLEX Plus Rat IL-13 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of IL-13 in rat serum plasma and cell culture supernatants The kit includes a plate coated with a rat IL-13 specific antibody a ruthenium labeled rat IL-13 detection antibody calibrators and buffers for 96 tests The ECL signal produced is proportional to the amount of IL-13 present in the sample This assay has high sensitivity wide dynamic range and excellent performance in complex biological matrices The kit provides reagents for the quantitative measurement of rat IL-13 in approximately 3 5 hours
Catalog No. 50-000-78343