Learn More
Meso Scale Discovery V-PLEX Rat IL-13 Kit
SDP
Supplier:  Meso Scale Discovery K153ODD4
Encompass
The Meso Scale Discovery V-PLEX Rat IL-13 Kit is a highly sensitive and quantitative immunoassay for the detection of rat interleukin 13 (IL-13) in cell culture supernatants serum plasma and other biological fluids This electrochemiluminescent assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide superior sensitivity a wide dynamic range and requires just 50 L of sample The V-PLEX Rat IL-13 Kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and assay diluent The kit can accurately quantify rat IL-13 concentrations from 1 53 to 2500 pg/mL with an inter-assay CV of 12%
Catalog No. 50-000-82255
Only null left
Add to Cart
Request a Quote