Learn More
Meso Scale Discovery V-PLEX Rat KC/GRO Kit
SDP
Supplier:  Meso Scale Discovery K153QTD1
Encompass
The V-PLEX Rat KC/GRO Kit from Meso Scale Discovery is a multiplex immunoassay kit for the quantitative measurement of rat keratinocyte chemoattractant (KC) and rat growth-related oncogene (GRO) in serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The electrochemiluminescent assay has a sensitivity of 1 5 pg/mL for KC and 0 8 pg/mL for GRO and can detect KC and GRO levels between 1 5-1500 pg/mL and 0 8-800 pg/mL respectively The assay takes approximately 3 hours to complete
Catalog No. 50-000-81172