Learn More
Meso Scale Discovery V-PLEX Viral Panel 1 Human Kit, 5 plates, 96-well format, multiplex assay, IL-1β, IL-6, IL-8, TNF-α, sub- to low pg/mL detection limits, research use only
SDP
Supplier:  Meso Scale Discovery K15345D2
Encompass
V-PLEX Viral Panel 1 Human Kit, 5 plates, 96-well format, multiplex assay, IL-1β, IL-6, IL-8, TNF-α, sub- to low pg/mL detection limits, research use only
Catalog No. NC2890179