Learn More
Eurofins Discoverx ChemiScreen V1A Vasopression Receptor Stable Cell Line, 2 vials. (HTS059C)
Supplier:  Eurofins Discoverx HTS059C
Encompass
The ChemiScreen V1A Vasopressin Receptor Stable Cell Line is an engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. vasopressin) based activation of AVPR1A GPCR activity via detection of increasing levels of intracellular calcium. The included cell line overexpresses AVPR1A alongside endogenous high levels of the promiscuous G protein, Gα15, coupling the receptor to the calcium signaling pathway. Upon activation of AVPR1A, the resulting calcium activity can be fluorescently assayed. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, visit www.discoverx.com/targets/gpcr-target-biology.
Catalog No. 50-200-5771
Only null left
Add to Cart
Request a Quote