Learn More
Cytek Biosciences Inc cFluor® V450 Anti-Human CD45RA (HI100)
Supplier:  Cytek Biosciences Inc R720122
Encompass_Preferred
The HI100 monoclonal antibody binds to the 220 kDa isoform of the human leukocyte common antigen. CD45RA is an isoform of CD45 due to alternate splicing1. CD45RA is expressed on naïve or resting CD4+ and CD8+ T cells, as well as on a subset of activated T cells2,3. The associated tyrosine phosphatase activity regulates signal transduction processes during hematopoiesis and immune responses4.The antibody was conjugated to a fluorophore and purified by affinity chromatography.
Catalog No. NC2823283