Learn More
Integra Biosciences Corp 300 µl Standard 5 Viaflo 96 Racks of 96 Tips
SDP
Supplier:  Integra Biosciences Corp 6435/DEL
Encompass
300 i1/4l, 5 racks of 96 tips By designing our GripTips in tandem with VIaFLO pipettes, we have created a unique, integrated system that offers the securest pipette tip connection. Without hammering, the GripTips literally snap onto the pipette's tip fitting, which prevents tips from falling off. Reduced tip attachment and ejection forces for comfortable, stress-free pipetting. Precise and consistent tip seal ensures that all tips on a multichannel pipette, including VIaFLO 96 and VIaFLO 384, are precisely at an identical height for reliable touch offs and consistent pipetting results.
Catalog No. NC0207862
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
View Alternate Product