Learn More
Medchemexpress LLC Va-k-14 hydrochloride | 1171341-19-7 | 99.0% | 25 MG

Supplier: Medchemexpress LLC HY12297425MG
VA-K-14 hydrochloride is a research-grade small molecule that antagonizes the thyroid-stimulating hormone receptor (TSHR). It is supplied as the hydrochloride salt in solid form and is intended for laboratory research use only.
- Specific TSHR antagonist with reported IC50 = 12.3 μM.
- Hydrochloride salt and solid physical form.
- Molecular weight 341.86 g·mol⁻¹; formula C18H16ClN3S.
- High purity (~99.0%) suitable for in vitro studies.
- Available in small research pack sizes such as 25 mg.
- For laboratory research only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.