Learn More
Medchemexpress LLC HY-111200 5mg , Vabicaserin hydrochloride CAS:887258-94-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1112005MG
Encompass_Preferred
HY-111200 5mg Vabicaserin hydrochloride CAS: 887258-94-8 Vabicaserin hydrochloride is a 5-hydroxytryptamine 2C (5-HT2C) receptor-selective agonist with an EC50 of 8 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0526