Learn More
Apexbio Technology LLC Vabicaserin hydrochloride 887258-94-8 5mg
SDP
Supplier:  Apexbio Technology LLC A46595
Encompass_Preferred
Vabicaserin hydrochloride (CAS 887258-94-8) is a selective agonist of the serotonin 5-HT2C receptor exhibiting an EC50 of 8 nM By engaging and modulating the 5-HT2C receptor-mediated signaling pathway this compound influences neurotransmitter release and neuronal excitability Vabicaserin hydrochloride has been widely utilized in preclinical models of neuropsychiatric disorders particularly in investigations pertaining to the pathophysiology and therapeutic intervention of anxiety and depression Its pharmacological profile makes it a valuable tool for studying serotonergic modulation of central nervous system function
Catalog No. 50-000-96329