Learn More
Medchemexpress LLC Vabicaserin hydrochloride | 887258-94-8 | 99.47% | 264.79 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY111200
Encompass_Preferred
Vabicaserin hydrochloride is a 5-hydroxytryptamine 2C (5-HT2C) receptor-selective agonist with an EC50 of 8 nM, intended for research use only. It shows high selectivity over other serotonergic, noradrenergic, and dopaminergic receptors, with potent agonistic activity in stimulating 5-HT2C receptor-coupled calcium mobilization.
  • 5-HT2C receptor-selective agonist
  • Potent and full agonist (EC50, 8 nM)
  • Stimulates 5-HT2C receptor-coupled calcium mobilization
  • Exhibits 5-HT2A receptor antagonism
  • Shows 5-HT2B antagonist or partial agonist activity
Catalog No. 50-003-46855