Learn More
Cayman Chemical VacquInol-1 10mg
SDP
Supplier:  Cayman Chemical 1632110 MG
Encompass_Preferred
An activator of MKK4-dependent macropinocytotic cell death in glioblastoma cells, inducing ATP depletion (IC50 = 3.14 µM at 1 day); orally bioavailable, substantially impairing the growth of engrafted glioblastoma cell tumors in mice
Catalog No. 50-297-2047