Learn More
Medchemexpress LLC Vacquinol-1 dihydrochloride (NSC13316 dihydrochloride) | 2309312-85-2 | 99.7% | 425.78 | 25 MG

Supplier: Medchemexpress LLC 25MGHY19345A
Vacquinol-1 dihydrochloride (NSC13316) is a MKK4-specific activator that activates MAPK pathways. It specifically induces human glioblastoma cell (GC) death, attenuates tumor progression, and prolongs survival in a glioblastoma multiforme (GBM) mouse model. This compound also induces apoptosis in hepatocellular carcinoma (HCC) cells.
- MKK4-specific activator
- Activates MAPK pathways
- Induces glioblastoma cell death
- Attenuates tumor progression in glioblastoma multiforme mouse models
- Prolongs survival in glioblastoma multiforme mouse models
- Induces apoptosis in hepatocellular carcinoma cells
- Purity of 99.7%
- Appearance: white to off-white solid
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.