Learn More
Medchemexpress LLC Vactosertib | 1352608-82-2 | 99.5% | 399.42 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY19928200MG
Encompass_Preferred
Vactosertib (EW-7197) is a potent, orally active, and ATP-competitive activin receptor-like kinase 5 (ALK5) inhibitor. It inhibits ALK2 and ALK4 at nanomolar concentrations and exhibits potent antimetastatic activity and anticancer effects. With a high purity of 99.53% and molecular weight of 399.42, this solid, off-white to yellow compound is a valuable tool for cancer research.
  • Potent and orally active ALK5 inhibitor
  • Inhibits ALK2 and ALK4
  • Shows antimetastatic and anticancer effects
  • Blocks TGFβ-induced phosphorylation of Smad2 or Smad3
  • Suppresses TGFβ-induced nuclear translocation of Smad2/3
  • Abrogates TGFβ1-induced tumor cell migration and invasion
  • Abolishes TGFβ1-induced effects on genes related to epithelial-to-mesenchymal transition
Catalog No. 50-004-01062