Learn More
Bellco Glass, Inc. Vactrap 2 Secondary Container Spill Basin, Safety Tray with Dividers, for use with Vactrap2 XXL Red 22in x 17in x 6in Vacuum Trap System or for General Lab Safety, Waste Storage, and Spill Containment, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800009
Encompass_Preferred
Vactrap2 is the world’s most advanced vacuum trap system. Ideal for the protection of vacuum lines and scientists alike, Vactrap2 safely contains waste from supernatant removal, chemical separation, and cell or tissue culture media aspiration. Best of all, Vactrap2 comes ready to use; simply attach to your vacuum and begin your work.Vactrap2 tubing extensions allow users to replace or extend the tubing connecting their Vactrap2 system to their vacuum. NEW Improved heavy-duty EZBio TPE V tubing to avoid kinking. Each extension includes a replacement inline vent filter. For more replacement vent filters, search ” vent filter”.Say goodbye to glass. Say hello to Vactrap2 .Specifications:Height: 6″ (153mm)Length: 22.5″ (572mm)Width: 17.5″ (431mm)Bin Material: PolypropyleneAutoclavable
Catalog No. 50-255-0746