Learn More
Medchemexpress LLC Vacuolin-1 | 351986-85-1 | 98.5% | 577.42 g/mol | C26H24IN7O | 100 MG
SDP
Supplier:  Medchemexpress LLC HY118630100MG
Encompass_Preferred
Vacuolin-1 is a cell-permeable small molecule inhibitor used in research to block Ca2+-dependent lysosomal exocytosis and inhibit PIKfyve, disrupting endosomal-lysosomal trafficking and late-stage autophagy. It is commonly used to study lysosomal maturation, autophagy, and cellular trafficking processes.
  • Selective inhibitor of PIKfyve and lysosomal exocytosis.
  • Cell-permeable small molecule suitable for cellular trafficking studies.
  • Impedes late-stage autophagy by disrupting lysosomal maturation.
  • Induces vacuole formation and can increase cellular enucleation.
  • Reported high purity for consistent experimental results.
Catalog No. 50-002-28059