Learn More
Med Vet International BD Vacutainer Plus SST™ Venous Blood Collection Tube, Clot Activator/Separator Gel Additive, 4 mL, Hemogard™ Closure Plastic Tube, 100/Box
Supplier:  Med Vet International 367977
Encompass_Preferred
Features Closure helps to protect lab personnel from contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube The rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact Clot Activator / Gel for Serum Separation
Catalog No. 50-304-3052