Learn More
Harrick Plasma Vacuum Gauge and Digital Meter 115V
SDP
Supplier:  Harrick Plasma PDCVCG
Encompass_Preferred
The PDC-VCG provides a vacuum gauge and digital meter as a stand-alone accessory and may be used with any of our plasma cleaners. The ability to monitor vacuum pressure during plasma processing is beneficial for process consistency. Vacuum gauge sensor connects to plasma cleaner door; Digital meter displays pressure range of 1 mTorr to 760 Torr; Size: 5″ H x 10″ W x 8″ D; Weight: 5 lb.
Catalog No. NC1462930