Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Vacuum Hose, 25 mm Diameter, 5 m Length

Supplier:  Waters Corp 700011035

Encompass_Preferred

The Waters vacuum hose provides a reliable solution for various laboratory vacuum applications. It is designed to ensure efficient and secure connections within experimental setups.

  • Equipped with a 25 mm diameter
  • Measures 5 m in length

Catalog No. 50-239-2467


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.