Learn More
Chemglass Life Sciences Vacuum Trap, Complete, Trap Bottle 32mm OD x 200mm Overall Length, 10mm OD Side and Inner Tubes, 29/42 Jt.
Supplier: Chemglass Life Sciences CG451401
Vacuum Trap, Complete, Trap Bottle 32mm OD x 200mm Overall Length, 10mm OD Side and Inner Tubes, 29/42 Jt. Two piece vacuum trap connected with an inner and outer joint to facilitate cleaning.