Learn More
Medchemexpress LLC Vadastuximab 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP993295MG
Encompass_Preferred
Vadastuximab (Anti-Human CD33 Recombinant Antibody) is a humanized monoclonal antibody targeting CD33 Vadastuximab can be used to synthesize an ADC compound Vadastuximab talirine[1]
Catalog No. 50-003-23974