Learn More
Selleck Chemical LLC Vafidemstat S9894-100MG
SDP
Supplier:  Selleck Chemical LLC S9894100MG
Encompass_Preferred
Vafidemstat(ORY-2001) is a potent selective brain-penetrant dual inhibitor of lysine-specific histone demethylase (LSD1 KDM1A) and MAO-B with IC50 values of 105 nM and 58 nM respectively It shows potential for treating memory deficits and behavioral alterations in neurodegenerative and psychiatric diseases
Catalog No. 50-000-23802