Learn More
Med Vet International VAGNA/SHEEP/GAT/ARTFCAL/KT/8
Supplier:  Med Vet International JOR125G
Encompass_Preferred
Sheep/Goat Artificial Vagina Kit - 8 Features Straight black rubber tube Latex liner Latex collection cone 15ml centrifuge tube 2 rubber bands for securing liner
Catalog No. 50-004-95738