Learn More
Medchemexpress LLC Val-Ala-PAB-MMAE | 1912408-92-4 | 98.9% | 1037.33 | C55H88N8O11 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15303225MG
Encompass_Preferred
Val-Ala-PAB-MMAE is a drug-linker conjugate for antibody-drug conjugate (ADC) research that couples a cleavable Val-Ala-PAB peptide linker to the cytotoxic payload monomethyl auristatin E (MMAE). It is supplied as a solid for conjugation and ADC synthesis.
  • Cleavable Val-Ala-PAB peptide linker for intracellular release.
  • Monomethyl auristatin E (MMAE) payload; effective tubulin inhibitor.
  • High purity suitable for research applications (98.9%).
  • White to off-white solid for convenient handling during conjugation.
  • Recommended storage at 4°C and protect from light.
Catalog No. 50-002-04347