Learn More
Medchemexpress LLC Val-Ala-PABC-Exatecan | 2845164-91-0 | 99.9% | 754.80 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1470955MG
Encompass_Preferred
Val-Ala-PABC-Exatecan is a Drug-Linker Conjugate for ADC, composed of a cleavable Tesirine linker and Exatecan, a topoisomerase I inhibitor. This product can be utilized for ADC molecule synthesis, such as Mal-PEGn-amide-va-Exatecan.
  • Consists of a cleavable Tesirine linker
  • Includes Exatecan, a topoisomerase I inhibitor
  • Can be used for ADC molecule synthesis
  • For research use only
Catalog No. 50-003-62193