Learn More
Medchemexpress LLC Val-ala-pabc-exatecan | 2845164-91-0 | 99.9% | 754.80 g·mol⁻1 | C40H43FN6O8 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY147095A50MG
Encompass_Preferred
Val-Ala-PABC-Exatecan is a drug-linker conjugate used in antibody-drug conjugate (ADC) synthesis that links a cleavable Val-Ala-PABC peptide linker to the topoisomerase I inhibitor exatecan. It is intended as a building block for ADC construction and linker-payload optimization studies.
  • Cleavable Val-Ala-PABC linker enabling protease-mediated payload release.
  • Exatecan payload; a topoisomerase I inhibitor.
  • High purity (99.9%) with characterized molecular weight and formula.
  • Suitable for ADC synthesis and linker-payload optimization studies.
  • Available in small research pack sizes, including 50 MG.
Catalog No. 50-002-02569