Learn More
Medchemexpress LLC Val-ala-pabc-exatecan | 2845164-91-0 | C40H43FN6O8 | 10MG
SDP
Supplier:  Medchemexpress LLC HY14709510MG
Encompass_Preferred
Val-Ala-PABC-Exatecan is a cleavable drug-linker conjugate that pairs a Val-Ala-PABC peptide linker with the topoisomerase I inhibitor exatecan. It is intended for incorporation into antibody-drug conjugates (ADCs) to enable enzymatic release of the cytotoxic payload inside target cells. Supplied as a solid with high analytical purity, it supports conjugation workflows and characterization by HPLC/LC-MS.
  • Cleavable Val-Ala-PABC linker enables enzymatic release of payload.
  • Includes exatecan, a potent topoisomerase I inhibitor.
  • High analytical purity suitable for conjugation and characterization.
  • Supplied as a stable solid for handling and storage.
  • Designed for ADC construction and linker-payload optimization.
Catalog No. 50-002-58747