Learn More
Medchemexpress LLC Val-Cit-amide-Cbz-N(Me)-Maytansine | 1628543-59-8 | 97.9% | 1140.71 g/mol | C55H78ClN9O15 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15689625MG
Encompass_Preferred
Val-Cit-amide-Cbz-N(Me)-Maytansine is a maytansine-based payload intermediate intended for research use in antibody-drug conjugate (ADC) synthesis and bioconjugation. It contains a valine-citrulline (Val-Cit) cleavable dipeptide linker and a Cbz-protected N-methyl maytansine moiety, provided as a powder for laboratory conjugation workflows.
  • Val-Cit cleavable dipeptide linker for cathepsin B-mediated release.
  • Cbz-protected N-methyl maytansine payload compatible with common conjugation chemistries.
  • High purity suitable for research applications.
  • Powder form with defined storage conditions for long-term stability.
  • Suitable as an intermediate for ADC construction and payload development.
Catalog No. 50-002-04423