Learn More
Medchemexpress LLC Val-cit-amide-ph-maytansine | 98.0% | 1055.61 | C51H71ClN8O14 | 10 MG

Supplier: Medchemexpress LLC HY15689710MG
Val-Cit-amide-Ph-Maytansine is a maytansine-derived small molecule intended for use as an antibody-drug conjugate (ADC) payload or linker component. It is supplied as a powder for formulation and conjugation studies and includes solubility and storage guidance for research handling.
- Val-cit-amide-ph-maytansine is a maytansine-derived ADC linker-payload.
- Purity is approximately 98.01%.
- Molecular weight 1055.61 and formula C51H71ClN8O14.
- Supplied as a powder with recommended storage at -20°C (long term) and 4°C (short term).
- Highly soluble in DMSO (100 mg/mL) and compatible with recommended in vivo formulations.
- Available in small research sizes (1 mg, 5 mg, 10 mg, 25 mg, 50 mg, larger sizes by quote).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.