Learn More
Medchemexpress LLC Val-cit-amide-ph-maytansine | 98.0% | 1055.61 | C51H71ClN8O14 | 25 MG

Supplier: Medchemexpress LLC HY15689725MG
Val-Cit-amide-Ph-Maytansine is a maytansine-derived payload used in antibody-drug conjugate research; it binds the hepatocyte growth factor receptor c-Met and is supplied for research use only.
- Purity: 98.01%.
- Molecular formula: C51H71ClN8O14.
- Molecular weight: 1055.61 g·mol⁻¹.
- Appearance: white to off-white solid.
- Solubility: soluble in DMSO (100 mg/mL); protocols available for in vivo formulation.
- Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
- Pack sizes: available in 1 mg-50 mg in stock; larger sizes by quote.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.