Learn More
Medchemexpress LLC Val-cit-amide-ph-maytansine | 98.0% | 1055.61 | C51H71ClN8O14 | 5 MG

Supplier: Medchemexpress LLC HY1568975MG
Val-cit-amide-ph-maytansine is a maytansine-based drug-linker designed for incorporation into antibody-drug conjugates (ADCs). It contains a valine-citrulline (Val-Cit) cleavable dipeptide linker connected to a maytansine payload via an amide-phenyl spacer, providing a protease-cleavable release mechanism for targeted delivery.
- intended for synthesis of antibody-drug conjugates and drug-linker research
- molecular weight 1055.61
- chemical formula C51H71ClN8O14 (product page) - alternative formula reported in datasheet
- purity reported as 98.01%
- physical form: powder; available in multiple vial sizes (1 mg-200 mg)
- storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years); in solution -80 °C (up to 6 months)
- solubility: DMSO 100 mg/mL (in vitro); in vivo vehicle formulations reported with solubility ≥ 2.5 mg/mL
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.