Learn More
Medchemexpress LLC Val-Cit-PAB-Exatecan | 2227350-99-2 | 99.8% | 840.90 g/mol | C43H49FN8O9 | 5 MG

Supplier: Medchemexpress LLC HY153031A5MG
Val-Cit-PAB-Exatecan is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links an exatecan payload to a cathepsin-cleavable Val-Cit-PAB linker, enabling intracellular release of the cytotoxic agent after lysosomal cleavage.
- Purity 99.8%.
- Molecular weight 840.90 g/mol.
- Chemical formula C43H49FN8O9.
- CAS number 2227350-99-2.
- Appearance light yellow to yellow solid.
- Storage -20°C, sealed, away from moisture and light; in solvent: -80°C for 6 months, -20°C for 1 month.
- Package available as 5 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.