Learn More
Medchemexpress LLC Val-Cit-PAB-Exatecan | 2227350-99-2 | 99.8% | 840.90 g/mol | C43H49FN8O9 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15303150MG
Encompass_Preferred
Val-Cit-PAB-Exatecan is a linker-drug conjugate composed of the topoisomerase I inhibitor exatecan (DX-8951) linked via a cathepsin-cleavable valine-citrulline p-aminobenzyl carbamate (Val-Cit-PAB) linker, supplied for research use in antibody-drug-conjugate development.
  • High purity suitable for research applications.
  • Cathepsin-cleavable Val-Cit-PAB linker for intracellular payload release.
  • Intended for antibody-drug conjugate synthesis and ADC research.
  • Provided as a solid, light yellow to yellow, with analytical data (H NMR, LCMS, RP-HPLC) available.
  • Available in multiple milligram sizes for small-scale conjugation work.
Catalog No. 50-002-01977