Learn More
Medchemexpress LLC Val-Cit-PAB-exatecan | 99.8% | 840.90 Da | C43H49FN8O9 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY15303125MG
Encompass_Preferred
Val-Cit-PAB-Exatecan is an antibody-drug conjugate (ADC) agent-linker conjugate composed of an exatecan payload (a DNA topoisomerase I inhibitor) attached via a cathepsin-cleavable valine-citrulline para-aminobenzyl (Val-Cit-PAB) linker. It is supplied as a research reagent intended for ADC synthesis and linker-payload development.
  • Cathepsin-cleavable Val-Cit-PAB linker
  • Exatecan payload (topoisomerase I inhibitor)
  • High purity suitable for research applications
  • Available in multiple package sizes, including 25 MG
  • Useful for ADC linker-payload conjugation studies
Catalog No. 50-002-02245