Learn More
Medchemexpress LLC Val-Cit-PAB-Exatecan TFA | 2928571-44-0 | 99.7% | 954.92 | C45H50F4N8O11 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY153031A25MG
Encompass_Preferred
Val-Cit-PAB-Exatecan TFA is an agent-linker conjugate for antibody-drug conjugates, composed of the topoisomerase I inhibitor Exatecan (DX-8951) linked via a cathepsin-cleavable Val-Cit-PAB linker. It is supplied for research use in multiple package sizes, including 25 mg.
  • Agent-linker conjugate for antibody-drug conjugates.
  • Composed of Exatecan and a cathepsin-cleavable val-cit-pab linker.
  • High purity (99.73%) as listed by manufacturer.
  • Molecular weight 954.92 (TFA salt form).
  • Available in multiple package sizes, including 25 mg.
Catalog No. 50-002-02246