Learn More
Medchemexpress LLC Val-Cit-PAB-MMAF sodium | 863971-56-6 | 98.4% | 1159.39 | C58H91N10NaO13 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY4417710MG
Encompass_Preferred
Val-Cit-PAB-MMAF sodium is an antibody-drug conjugate (ADC) agent-linker conjugate that couples the Val-Cit-PAB peptide linker to the tubulin inhibitor MMAF. It is supplied as a sodium salt, provided as a solid, and is intended for ADC research and linker-payload development.
  • High purity (98.4%).
  • Supplied as a solid; soluble in DMSO at 50 mg/mL (requires ultrasonic treatment).
  • Stable when stored under nitrogen: 4°C for solid; in solution, -80°C for 6 months, -20°C for 1 month.
  • Available in small research sizes, e.g., 10 mg.
  • Contains the Val-Cit-PAB peptide linker conjugated to the MMAF payload.
Catalog No. 50-002-02915