Learn More
Medchemexpress LLC Val-cit-pab-mmaf (sodium) | 98.4% | 1159.39 g/mol | C58H91N10NaO13 | 100 MG

Supplier: Medchemexpress LLC HY44177100MG
Val-Cit-PAB-MMAF sodium is a drug-linker conjugate used in antibody-drug conjugate (ADC) research that combines the cathepsin-cleavable peptide linker Val-Cit-PAB with monomethyl auristatin F (MMAF) as the cytotoxic payload. Intended for research use only.
- Drug-linker conjugate for ADC research.
- Purity approximately 98.4%.
- Molecular weight 1159.39 g/mol.
- Formula C58H91N10NaO13.
- Appearance: solid, off-white to light yellow.
- Storage: store at 4°C under nitrogen; in solvent, -80°C (6 months) or -20°C (1 month) when under nitrogen.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.