Learn More
Medchemexpress LLC Val-cit-PAB-OH | 159857-79-1 | MFCD 22381209 | 99.6% | 379.45 g/mol | C18H29N5O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HY123625G
Encompass_Preferred
Val-cit-PAB-OH is a protease-cleavable antibody-drug conjugate (ADC) linker that combines a valine-citrulline dipeptide with a para-aminobenzyl (PAB) self-immolative spacer. It is designed to release cytotoxic payloads inside target cells following lysosomal protease cleavage, and is supplied as a high-purity solid for conjugation chemistry and linker optimization.
  • Protease-cleavable Val-Cit dipeptide for intracellular release.
  • Para-aminobenzyl self-immolative spacer enabling clean payload liberation.
  • High reported purity suitable for conjugation workflows.
  • Well-characterized chemical formula and molecular weight for analytical planning.
  • Available in multiple package sizes to support discovery and scale-up.
Catalog No. 50-002-04051