Learn More
Medchemexpress LLC Valacyclovir-d8 (hydrochloride) | 1279033-32-7 | 98.0% | 368.85 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17425AS5MG
Encompass_Preferred
Valacyclovir-d8 hydrochloride is a deuterium-labeled, orally active antiviral agent. It is effective against herpes simplex, herpes zoster, and herpes B, and acts as a proagent of Aciclovir.
  • Deuterium labeled for use as a tracer or internal standard for quantitative analysis.
  • Orally active antiviral agent effective for herpes simplex, herpes zoster, and herpes B.
  • Inhibits HSV-1 W with an IC50 of 2.9 μg/ml.
  • Proagent of Aciclovir.
Catalog No. 50-003-84273