Learn More
Medchemexpress LLC Valacyclovir (hydrochloride) | 124832-27-5 | 99.02% | 360.80 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY17425A
Encompass_Preferred
Valacyclovir hydrochloride (Valaciclovir hydrochloride) is an orally active antiviral agent effective against herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W (50=2.9 μg/ml) and is a prodrug of Aciclovir (HY-17422). This product is intended for research and analytical applications.
  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 W (50=2.9 μg/ml)
  • Prodrug of aciclovir
  • Intended for research and analytical applications
Catalog No. 50-003-84021