Learn More
Medchemexpress LLC Valacyclovir hydrochloride | 124832-27-5 | 99.02% | 360.80 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY17425AR50MG
Encompass_Preferred
Valacyclovir (hydrochloride) (Standard) is the analytical standard of Valacyclovir (hydrochloride), an orally active antiviral agent for herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W (50=2.9 μg/ml) and is a proagent of Aciclovir. This product is intended for research and analytical applications, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard
  • Orally active antiviral agent
  • Effective against herpes simplex, herpes zoster, and herpes B
  • Inhibits HSV-1 W (50=2.9 μg/ml)
  • Proagent of Aciclovir
  • Suitable for research and analytical applications
  • Used in HPLC, GC, and MS experiments
Catalog No. 50-003-85644