Learn More
Medchemexpress LLC Valacyclovir hydrochloride (Standard) | 124832-27-5 | 99.0% | 360.80 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY17425AR5MG
Encompass_Preferred
Valacyclovir (hydrochloride) (Standard) is the analytical standard of Valacyclovir (hydrochloride). This product is intended for research and analytical applications. Valacyclovir hydrochloride (Valaciclovir hydrochloride) is an orally active antiviral agent for herpes simplex, herpes zoster, and herpes B. It inhibits HSV-1 W (50=2.9 μg/ml). Valacyclovir hydrochloride is a proagent of Aciclovir (HY-17422).
  • The compound is the grade of analytical standard, which is the reference standard supplied assay.
  • It is commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-003-84605