Learn More
Medchemexpress LLC Valategrast (R-411 free base) | 220847-86-9 | 97.1% | C30H32Cl3N3O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY14190
Encompass_Preferred
Valategrast (R-411 free base) is a potent and orally active integrin α4β1 (VLA-4) and α4β7 dual antagonist. It has potential for Chronic obstructive pulmonary disease (COPD) and asthma treatment.
  • Potent and orally active integrin α4β1 (VLA-4) and α4β7 dual antagonist
  • Potential for chronic obstructive pulmonary disease (COPD) and asthma treatment
  • Contains a common l-phenylalanine-N-aroyl motif
  • Carboxylic acid is responsible for binding to the metal ion in the metal-ion-dependent adhesion site (MIDAS)
Catalog No. 50-003-52620